Order Entry
Portugal
ContactUsLinkComponent
C18H x 3µm particle size x 120Å pore size, 50mm length x 300µm ID, PEEKsil ProteCol HPLC Capillary Column 1 * 1 items
Catalog # SGEA2C182-03L30K
C18H x 3µm particle size x 120Å pore size, 50mm length x 300µm ID, PEEKsil ProteCol HPLC Capillary Column 1 * 1 items
Catalog # SGEA2C182-03L30K
Frequently Bought Together