Order Entry
Portugal
ContactUsLinkComponent
HILIC HC x 5µm particle size x 120Å pore size, 100mm length x 2.1mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2HL9-05M02S
undefined
HILIC HC x 5µm particle size x 120Å pore size, 100mm length x 2.1mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2HL9-05M02S
Frequently Bought Together