Order Entry
Portugal
ContactUsLinkComponent
Cyano x 5µm particle size x 120Å pore size, 150mm length x 4.6mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2CN-05N46S
Cyano x 5µm particle size x 120Å pore size, 150mm length x 4.6mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2CN-05N46S
Frequently Bought Together