Order Entry
Portugal
ContactUsLinkComponent
Cyano x 5µm particle size x 120Å pore size, 150mm length x 3.0mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2CN-05N03S
Cyano x 5µm particle size x 120Å pore size, 150mm length x 3.0mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2CN-05N03S
Frequently Bought Together