Order Entry
Portugal
ContactUsLinkComponent
Cyano x 5µm particle size x 120Å pore size, 100mm length x 2.1mm ID, PEEK-coated ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2CN-05M02P
Cyano x 5µm particle size x 120Å pore size, 100mm length x 2.1mm ID, PEEK-coated ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2CN-05M02P
Frequently Bought Together