Order Entry
Portugal
ContactUsLinkComponent
C8H x 5µm particle size x 120Å pore size, 150mm length x 1.0mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2C83-05N01S
C8H x 5µm particle size x 120Å pore size, 150mm length x 1.0mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2C83-05N01S
Frequently Bought Together