Order Entry
Portugal
ContactUsLinkComponent
C8H x 3µm particle size x 120Å pore size, 100mm length x 150µm ID, PEEKsil ProteCol HPLC Capillary Column 1 * 1 items
Catalog # SGEA2C83-03M15K
C8H x 3µm particle size x 120Å pore size, 100mm length x 150µm ID, PEEKsil ProteCol HPLC Capillary Column 1 * 1 items
Catalog # SGEA2C83-03M15K
Frequently Bought Together