Order Entry
Portugal
ContactUsLinkComponent
Elastase is a serine protease that preferentially cleaves at the C-terminus of alanine, valine, serine, glycine, leucine or isoleuci ne. Elastase has a unique capability of digesting elastin. This enzyme can be used alone or in combination with other 1 * 5 mg
Catalog # PROMV1891
Supplier:  Promega
undefined
Elastase is a serine protease that preferentially cleaves at the C-terminus of alanine, valine, serine, glycine, leucine or isoleuci ne. Elastase has a unique capability of digesting elastin. This enzyme can be used alone or in combination with other 1 * 5 mg
Catalog # PROMV1891
Supplier:  Promega
Frequently Bought Together