O-rings
O-rings, viton, type KF, DN 25
Catalog # ILMV701173
Supplier: Welch by Gardner Denver
Jump to:
Product Details & Documents
O-rings, Type KF made of Perbunan® or Viton®.
Specifications
- Description:O-rings, viton, type KF, DN 25
- Seq:04