O-rings
O-rings, perbunan, type KF, DN 16
Catalog # ILMV701161
Supplier: Welch by Gardner Denver
Jump to:
Product Details & Documents
O-rings, Type KF made of Perbunan® or Viton®.
Specifications
- Description:O-rings, perbunan, type KF, DN 16
- Seq:01