Thymol blue
Glass bottle, 25 g
Catalog # TCIAT0235-25G
Supplier: TCI
Jump to:
Product Details & Documents
Synonyms: Thymolsulphonphthalein, 3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
- CAS Number:76-61-9
- Formula:C₂₇H₃₀O₅S
- MDL Number:MFCD00005869
- Molecular Weight:466.6 g/mol
- Merck Index:14,09400
- Melting Pt:221 °C
- Storage Temperature:Ambient
- EINECS:200-973-3
Documents
Specifications
Specification Test Results
- Appearance:Yellow to Brown to Dark green powder to crystaline
- Absorbance:min. 0.270 (10mg/L, pH 7.6, 431.0 to 437.0 nm)
- Solubility in Sodium hydroxide solution:almost transparency
- Transition interval (pH):(red)1.2 to 2.8 (yellow), (yellow)8.0 to 9.6 (blue)
Specifications
- Pack type:Glass bottle
- Pk:25 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



