Order Entry
Portugal
Orders LinkContactUsLinkComponent
 

Thymol blue

Glass bottle, 1 g
Catalog # TCIAT0235-1G
Supplier:   TCI
Thymol blue
Catalog # TCIAT0235-1G
Supplier:  TCI
CAS Number:  76-61-9
Thymol blue
Thymol blue

Glass bottle, 1 g

CAS Number:  76-61-9
Price
25,00 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  Thymolsulphonphthalein, 3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
  • CAS Number:76-61-9
  • Formula:C₂₇H₃₀O₅S
  • MDL Number:MFCD00005869
  • Molecular Weight:466.6 g/mol
  • Merck Index:14,09400
  • Melting Pt:221 °C
  • Storage Temperature:Ambient
  • EINECS:200-973-3
Specifications

Specification Test Results

  • Appearance:
    Yellow to Brown to Dark green powder to crystaline
  • Absorbance:
    min. 0.270 (10mg/L, pH 7.6, 431.0 to 437.0 nm)
  • Solubility in Sodium hydroxide solution:
    almost transparency
  • Transition interval (pH):
    (red)1.2 to 2.8 (yellow), (yellow)8.0 to 9.6 (blue)

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g

Specifications