Order Entry
Portugal
Orders LinkContactUsLinkComponent
dsDNA System, QuantiFluor®
QuantiFluor® dsDNA System, sufficient reagents for 2000 assays at a 200 µl scale, 1 mL
  PROME2670
 :   Promega
dsDNA System, QuantiFluor®
  PROME2670
 :  Promega
 :  
dsDNA System, QuantiFluor®
dsDNA System, QuantiFluor®
QuantiFluor® dsDNA System, sufficient reagents for 2000 assays at a 200 µl scale, 1 mL
 :  
639,00 €
1 * 1 mL
The QuantiFluor® dsDNA System contains a fluorescent DNA-binding dye that enables sensitive quantitation of small amounts of double-stranded DNA (dsDNA) in solution. The system is suitable for use in many molecular biology applications, including next-gen sequencing and other techniques where accurate DNA quantitation is important.
  • Specific quantitation of dsDNA
  • More sensitive than other methods for low-concentration samples
QuantiFluor® dsDNA dye shows minimal binding to single stranded DNA (ssDNA), RNA, protein and interfering compounds, allowing specific quantitation of dsDNA.
 : Kit includes QuantiFluor® dsDNA dye (1×1 ml), Lambda DNA standard (1×100 µg) and 20X TE buffer, pH 7,5 (1×25 ml).
  • Pk:
    1 mL
  • Description:
    QuantiFluor® dsDNA System, sufficient reagents for 2000 assays at a 200 µl scale