Order Entry
Portugal
ContactUsLinkComponent
RECOMBINANT/NATIVE PROTEIN, FCGR2A (HUMAN) RECOMBINANT PROTEIN, HUMAN FCGR2A (P12318, GLN 34 - GLY 216) PARTIAL RECOMBINANT PROTEIN EXPRESSED IN HEK293 CELLS. 1 * 20 µG
Catalog # ABNOP3684
Supplier:  Abnova
RECOMBINANT/NATIVE PROTEIN, FCGR2A (HUMAN) RECOMBINANT PROTEIN, HUMAN FCGR2A (P12318, GLN 34 - GLY 216) PARTIAL RECOMBINANT PROTEIN EXPRESSED IN HEK293 CELLS. 1 * 20 µG
Catalog # ABNOP3684
Supplier:  Abnova