Order Entry
Portugal
ContactUsLinkComponent
KIT, GLYCEROL CELL-BASED ASSAY KIT, GLYCEROL CELL-BASED ASSAY KIT IS A COLORIMETRIC METHOD FOR QUANTIFYING GLYCEROL. 1 * 1 mL
Catalog # ABNOKA1317
Supplier:  Abnova
KIT, GLYCEROL CELL-BASED ASSAY KIT, GLYCEROL CELL-BASED ASSAY KIT IS A COLORIMETRIC METHOD FOR QUANTIFYING GLYCEROL. 1 * 1 mL
Catalog # ABNOKA1317
Supplier:  Abnova