Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Portugal
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
KIT, TTR (HUMAN) ELISA KIT, TTR (HUMAN) ELISA KIT IS A SANDWICH ENZYME IMMUNOASSAY FOR THE QUANTITATIVE MEASUREMENT OF HUMAN TTR. 1 * 1 KIT
Catalog # ABNOKA0495
Supplier:
Abnova
Expand image
KIT, TTR (HUMAN) ELISA KIT, TTR (HUMAN) ELISA KIT IS A SANDWICH ENZYME IMMUNOASSAY FOR THE QUANTITATIVE MEASUREMENT OF HUMAN TTR. 1 * 1 KIT
Catalog # ABNOKA0495
Supplier:
Abnova
Related Information
Documentation
Additional Product Details
Literature reference - PubMed ID 11693529
Literature reference - PubMed ID 11989633
Literature reference - PubMed ID 16316355
Literature reference - PubMed ID 22930475
Literature reference - PubMed ID 25728271
Datasheet
sds_ABNOKA0495.pdf
Documentation
Frequently Bought Together
Transfer pipettes
Supplier: RATIOLAB
VWR® Premium Line, Serological Pipettes
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 136x136 mm
Supplier: VWR Collection
Eppendorf® Safe-Lock Tubes, 0,5 ml - Microtubes
Supplier: EPPENDORF
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Sampling Spoons
Supplier: VWR Collection
VWR®, Microcentrifuge Tubes, High Performance
Supplier: VWR Collection
Serological pipettes, Stripette™
Supplier: Corning
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
VWR®, Petri Dish, PS
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Cell culture flasks, Corning®
Supplier: Corning
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
KIT, AFP (HUMAN) ELISA KIT, AFP (HUMAN) ELISA KIT IS A SANDWICH ENZYME IMMUNOASSAY FOR THE QUANTITATIVE MEASUREMENT OF HUMAN AFP IN PLASMA, SERUM, AND CELL CULTURE SUPERNATANTS. 1 * 1 KIT
Supplier: Abnova