Order Entry
Portugal
ContactUsLinkComponent
STIRRED BATH/CIRCULATOR, OPTIMA TX150 PROGRAMMABLE IMMERSION THERMOSTAT + ST26 STAINLESS STEEL TANK, 26L CAPACITY, INCL DRAIN TAP A ND PUMP. TEMP RANGE AMBIENT +5 TO 150C. 1 * 1 items
Catalog # 461-0708
Supplier:  GRANT INSTRUMENTS
STIRRED BATH/CIRCULATOR, OPTIMA TX150 PROGRAMMABLE IMMERSION THERMOSTAT + ST26 STAINLESS STEEL TANK, 26L CAPACITY, INCL DRAIN TAP A ND PUMP. TEMP RANGE AMBIENT +5 TO 150C. 1 * 1 items
Catalog # 461-0708
Supplier:  GRANT INSTRUMENTS
Frequently Bought Together