BILE SALTS 1 * 10 g
Catalog # SIALB8756-10G
Supplier: Merck
Frequently Bought Together
2-Propanol ≥99.5% (by GC), EMPLURA®, Supelco®
Supplier: Merck
Sodium dihydrogen phosphate dihydrate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Isooctane (Reag. Ph. Eur.) for analysis, ACS 1 * 2,5 L
Supplier: PanReac AppliChem
Dichloromethane stabilized with 20 ppm of amylene for pesticide analysis 1 * 2,5 L
Supplier: PanReac AppliChem
(±)-2-Butanol ≥99.5% (by GC), Puriss. p.a. Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
2-Propanol 99.6% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
2-Propanol ≥99.5% (by GC), Puriss., meets analytical specification of BP, Ph. Eur., USP, Riedel-de Haën™
Supplier: Honeywell Chemicals
Dimethyl sulphoxide 99.9% for spectroscopy
Supplier: Thermo Fisher Scientific
2,2,4-Trimethylpentane ≥99.5%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Lignin (dealkaline)
Supplier: TCI