Order Entry
Portugal
Orders LinkContactUsLinkComponent
Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer), Sigma-Aldrich®

Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer), Sigma-Aldrich®

TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer), 5 L
Catalog # SIAL93290-5L
Supplier:   Merck
Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer), Sigma-Aldrich®
Catalog # SIAL93290-5L
Supplier:  Merck
CAS Number:  
Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer), Sigma-Aldrich®

TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer), 5 L

CAS Number:  
Price
547,00 €
Log in for availability
1 * 5 L
Product Details & Documents
Specifications

Specifications

  • Pk:
    5 L
  • Description:
    TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer)

Specifications