LIPASE B, RECOMBINANT FROM CANDIDA ANT-A RCTICA, 9 U/MG 1 * 50 mg
SIAL62288-50MG-F
: Merck
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
2-Propanol ≥99.0%, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals