Order Entry
Portugal
ContactUsLinkComponent
6 COMPOSES AS,CD,MN,PB,SE,ZN DANS HNO3 5% As 1000mg/l Cd 100mg/l Mn 1000mg/l Pb 1000mg/l Se 1000mg/l Zn 100mg/l 1 * 100 mL
Catalog # CMPLA191.1K.5N.L1
6 COMPOSES AS,CD,MN,PB,SE,ZN DANS HNO3 5% As 1000mg/l Cd 100mg/l Mn 1000mg/l Pb 1000mg/l Se 1000mg/l Zn 100mg/l 1 * 100 mL
Catalog # CMPLA191.1K.5N.L1