Accessories
Base for separate media supply to enable suctioning of the medium directly from a separate storage container using a tube
Catalog # 613-3861
Supplier: HIRSCHMANN
Jump to:
Product Details & Documents
Documents
Specifications
- Description:Base for separate media supply to enable suctioning of the medium directly from a separate storage container using a tube
- Seq:13