Order Entry
Portugal
Orders LinkContactUsLinkComponent
Needles for Reacti Vap sample concentrator
Needles for Reacti Vap sample concentrator
Stainless steel PTFE coated needles, blunt end, length: 102 mm, 19, 102 mm
Catalog # HYPETS-18784
Needles for Reacti Vap sample concentrator
Catalog # HYPETS-18784
Needles for Reacti Vap sample concentrator
Stainless steel PTFE coated needles, blunt end, length: 102 mm, 19, 102 mm
Price
248,00 €
Log in for availability
1 * 9 unid.
Product Details & Documents
Specifications
  • Gauge:
    19
  • Description:
    Stainless steel PTFE coated needles, blunt end, length: 102 mm
  • Length:
    102 mm
  • Seq:
    031
Recomendações