Order Entry
Portugal
ContactUsLinkComponent
MICROTUBE THERMO-SHAKER WITH PSC24 BLOCK FOR 24 X 2.0 ML MICROTUBES, TEMPERATURE RANGE AMBIENT +5 TO 100C, SHAKING SPEED 250 TO 1400 RPM 1 * 1 items
Catalog # 444-1028
Supplier:  GRANT INSTRUMENTS
MICROTUBE THERMO-SHAKER WITH PSC24 BLOCK FOR 24 X 2.0 ML MICROTUBES, TEMPERATURE RANGE AMBIENT +5 TO 100C, SHAKING SPEED 250 TO 1400 RPM 1 * 1 items
Catalog # 444-1028
Supplier:  GRANT INSTRUMENTS
Frequently Bought Together