Order Entry
Portugal
Orders LinkContactUsLinkComponent
Washers, BOLA

Washers, BOLA

M 4 thread, 9 mm
Catálogo # BOHLH1126-14
Fornecedor:   Bohlender
Washers, BOLA
Catálogo # BOHLH1126-14
Fornecedor:  Bohlender
Washers, BOLA
Washers, BOLA

M 4 thread, 9 mm

Price
3,00 €
Iniciar Sessão para disponibilidade
1 * 1 unid.
Product Details & Documents

PTFE, white.

  • Temperature resistance: from −200 to +250 °C
  • Universal chemical resistance
  • FDA conform
  • Similar to DIN 125-1
Especificações
  • Ø:
    9 mm
  • Description:
    M 4 thread
Recomendações

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.