Order Entry
Portugal
ContactUsLinkComponent
33 to 650 rpm / 115/230 VAC / IP55 Digital display shows motor speed, from 5 to 100% Speed control via three-turn potentiometer 1 * 1 items
Catalog # ISMCMF0388
33 to 650 rpm / 115/230 VAC / IP55 Digital display shows motor speed, from 5 to 100% Speed control via three-turn potentiometer 1 * 1 items
Catalog # ISMCMF0388
Frequently Bought Together