Order Entry
Portugal
Orders LinkContactUsLinkComponent
Cyclohexanone, Sigma-Aldrich®

Cyclohexanone, Sigma-Aldrich®

10 kg
Catálogo # SIALW390909-10KG
Flammable
Irritant
Fornecedor:   Merck
Cyclohexanone, Sigma-Aldrich®
Catálogo # SIALW390909-10KG
Fornecedor:  Merck
Número CAS:  108-94-1
Cyclohexanone, Sigma-Aldrich®
Cyclohexanone, Sigma-Aldrich®

10 kg

Número CAS:  108-94-1
Price
218,00 €
Log in for availability
1 * 10 kg
Product Details & Documents
Synonyms:  Pimelic Ketone
  • CAS Number:108-94-1
  • Formula:C₆H₁₀(=O)
  • MDL Number:MFCD00001625
  • Molecular Weight:98.14 g/mol
  • Merck Index:13,02755
  • Boiling Pt:155.6 °C (1013 hPa)
  • Melting Pt:–31 °C
  • Density:0.9478 g/cm³ (20 °C)
  • Flash Pt:43 °C
  • UN:1915
  • ADR:3,III
  •  :M-Clarity™ quality level = MQ300
  • Storage Temperature:Ambient
  • EINECS:203-631-1
  • REACH:01-2119453616-35
Specifications

Specifications

  • Pk:
    10 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.