Order Entry
Portugal
Orders LinkContactUsLinkComponent
Vanillin, Sigma-Aldrich®

Vanillin, Sigma-Aldrich®

Glass bottle for solids, 1 kg
Catalog # SIALW310700-1KG-K
Irritant
Supplier:   Merck
Vanillin, Sigma-Aldrich®
Catalog # SIALW310700-1KG-K
Supplier:  Merck
CAS Number:  121-33-5
Vanillin, Sigma-Aldrich®
Vanillin, Sigma-Aldrich®

Glass bottle for solids, 1 kg

CAS Number:  121-33-5
Price
239,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  4-Hydroxy-3-methoxybenzaldehyde
  • CAS Number:121-33-5
  • Formula:C₈H₈O₃
  • MDL Number:MFCD00006942
  • Molecular Weight:152.15 g/mol
  • Merck Index:13,09998
  • Boiling Pt:285 °C (1013 hPa)
  • Melting Pt:∼82 °C
  • Density:1.056 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ300
  • Flash Pt:153 °C
  • Storage Temperature:Ambient
  • EINECS:204-465-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.