Order Entry
Portugal
Orders LinkContactUsLinkComponent
Silicon dioxide, Sigma-Aldrich®

Silicon dioxide, Sigma-Aldrich®

Glass bottle for solids, 250 g
Catalog # SIAL637246-250G
Supplier:   Merck
Silicon dioxide, Sigma-Aldrich®
Catalog # SIAL637246-250G
Supplier:  Merck
CAS Number:  7631-86-9
Silicon dioxide, Sigma-Aldrich®

Glass bottle for solids, 250 g

CAS Number:  7631-86-9
Price
918,00 €
Log in for availability
1 * 250 g
Product Details & Documents
Synonyms:  Silicon(IV) oxide, Silicium dioxide, Silicium (IV) oxide
  • CAS Number:7631-86-9
  • Formula:SiO₂
  • MDL Number:MFCD00011232
  • Molecular Weight:60.08 g/mol
  • Merck Index:14,08493
  • Boiling Pt:2230 °C (1013 hPa)
  • Melting Pt:1719 °C
  • Density:2.2 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ100
  • Storage Temperature:Ambient
  • EINECS:231-545-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.