GRUBBS CATALYST 2ND GENERATION 1 * 500 mg
Catalog # SIAL569747-500MG
Supplier: Merck
Frequently Bought Together
Methyltrichlorosilane for synthesis, Sigma-Aldrich®
Supplier: Merck
Heptafluorobutyric anhydride (HFAA), Supelco®
Supplier: Merck
1-Propanol ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethyl acetate ≥99.5% for HPLC
Supplier: Thermo Fisher Scientific
Methanol, extra dry 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
n-Hexane ≥97%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethyl acetate 99.6% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Benzyl alcohol 99%, pure
Supplier: Thermo Fisher Scientific
Hexane (mixture of isomers) ≥98.5%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Chloroform ≥99.5% stabilised
Supplier: Honeywell Chemicals
Diisopropyl ether stabilised for analysis
Supplier: Thermo Fisher Scientific
4,4'-Bis(dimethylamino)diphenylmethane 98%
Supplier: Thermo Fisher Scientific