Order Entry
Portugal
Orders LinkContactUsLinkComponent
Plasmid maxiprep system, PureYield™
PureYield™ plasmid Maxiprep start-up kit, 220 V, 50 preparations, with manifold, 4 elution devices and free vacuum pump, 1 SET
Catalog # PROMA6732
Supplier:   Promega
Plasmid maxiprep system, PureYield™
Catalog # PROMA6732
Supplier:  Promega
CAS Number:  
Plasmid maxiprep system, PureYield™
Plasmid maxiprep system, PureYield™
PureYield™ plasmid Maxiprep start-up kit, 220 V, 50 preparations, with manifold, 4 elution devices and free vacuum pump, 1 SET
CAS Number:  
Price
2 250,00 €
Log in for availability
1 * 1 SET
Product Details & Documents
The PureYield™ plasmid Maxiprep system is designed to isolate transfection quality plasmid DNA. The system provides a rapid method for purification up to 1 mg of plasmid DNA from a 250 ml bacterial culture.
  • Improved productivity: Vacuum protocol simplifies purification of multiple samples at one time
  • High purity and concentration of plasmid DNA gives proven performance in transfection, in vitro expression and other molecular biology applications
  • Simple protocol eliminates tedious high speed centrifugation, gravity drip columns, and post elution alcohol precipitation
  • PureYield™ membrane column allows purification of large amounts of plasmid DNA, exceeding the capabilities of other midiprep systems
The system incorporates a unique endotoxin removal wash designed to remove substantial amounts of protein, RNA and endotoxin contaminants from purified plasmid DNA. Removal of contaminants improves the robustness of sensitive applications such as eukaryotic transfection, in vitro transcription and coupled in vitro transcription/translation such as TnT® Quick Coupled transcription/translation system. Purification is achieved without isopropanol precipitation of purified plasmid DNA.
 : Kit components can be ordered separately.
 : The PureYield™ Maxiprep system includes PureYield™ Maxi binding columns, PureYield™ clearing columns, endotoxin removal wash, cell resuspension solution (CRA), cell lysis solution (CLA), neutralisation solution (NSB), column wash and nuclease-free water. The start-up kit includes Eluator™ vacuum elution device, PureYield™ plasmid Maxiprep system, welch vacuum pump and Vac-Man® laboratory vacuum manifold (20 sample capacity).
Specifications
Specifications
  • Pk:
    1 SET
  • Description:
    PureYield™ plasmid Maxiprep start-up kit, 220 V, 50 preparations, with manifold, 4 elution devices and free vacuum pump
Specifications
Accessories
Product Information
  • Catalog #
    Pk
    Availability
    Price
  • Vacuum elution device, Eluator™
    Vacuum elution device, Eluator™
    PROMA1071
    4
    Log in for availability
    1 * 4 unid. - 234,00 €