Order Entry
Portugal
Orders LinkContactUsLinkComponent
Multiple distributors for bottles with GLS 80 thread, BOLA

Multiple distributors for bottles with GLS 80 thread, BOLA

GLS 80, max. 12,7 mm, 4 × GL 18
Catalog # 201-0293
Supplier:   Bohlender
Multiple distributors for bottles with GLS 80 thread, BOLA
Catalog # 201-0293
Supplier:  Bohlender
Multiple distributors for bottles with GLS 80 thread, BOLA
Multiple distributors for bottles with GLS 80 thread, BOLA

GLS 80, max. 12,7 mm, 4 × GL 18

Price
126,00 €
Log in for availability
1 * 1 unid.
Product Details & Documents

PPS screw cap, blue, distributor body made from PTFE.

  • Very good chemical resistance
  • Temperature resistant from –200 to +250 °C
  • Suitable for vacuum applications

For the removal or filling of aggressive or pure liquids. For the connection of hoses, tubes and probes into vessels. Hoses with an external diameter of maximum 12,7 mm can be inserted through the stopcocks.

Specifications
  • Thread:
    GLS 80
  • Tube ext.Ø:
    max. 12,7 mm
  • Neck:
    4 × GL 18
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.