Order Entry
Portugal
ContactUsLinkComponent
Quickblue Capra hircus (goat). Complete PCR kit for sensitive and specific detection of the species indicated in food and other samp les. The kit contains the primer set, Taq polymerase, UNG enzyme as well as reference DNA. The protocol is in complia 1 * 10 Tests
Catalog # 710-0653
Supplier:  Q BIOANALYTIC
undefined
Quickblue Capra hircus (goat). Complete PCR kit for sensitive and specific detection of the species indicated in food and other samp les. The kit contains the primer set, Taq polymerase, UNG enzyme as well as reference DNA. The protocol is in complia 1 * 10 Tests
Catalog # 710-0653
Supplier:  Q BIOANALYTIC
Frequently Bought Together