Order Entry
Portugal
ContactUsLinkComponent
-86°C Upright Freezer. 382 litres. External Dimensions: 870 x 780 x 1975mm (wdh). Internal Dimensions: 620 x 515 x 1200mm (wdh). 10 0 % CFC Free refrigerant gas and insulation. Two inner doors. 1 * 1 items
Catalog # 471-0419
Supplier:  PHC Corporation
-86°C Upright Freezer. 382 litres. External Dimensions: 870 x 780 x 1975mm (wdh). Internal Dimensions: 620 x 515 x 1200mm (wdh). 10 0 % CFC Free refrigerant gas and insulation. Two inner doors. 1 * 1 items
Catalog # 471-0419
Supplier:  PHC Corporation
Frequently Bought Together