Order Entry
Portugal
ContactUsLinkComponent
Ph-combination electrode with plug head, glass shaft, ceramic diaphragm, electrolyte kcl 3 mol/l, iodine/iodide-reference system, sp here membrane, a-glass, length 120 mm, 12 mm , -5...+100 c, 0...14 ph 1 * 1 items
Catalog # 662-6088
Supplier:  SI Analytics
Ph-combination electrode with plug head, glass shaft, ceramic diaphragm, electrolyte kcl 3 mol/l, iodine/iodide-reference system, sp here membrane, a-glass, length 120 mm, 12 mm , -5...+100 c, 0...14 ph 1 * 1 items
Catalog # 662-6088
Supplier:  SI Analytics
Frequently Bought Together