Insert, volume 6mm, colour clear, material national scientific silanized glass, usable volume 300µl, conical base, polyspring, resid ual volume <1µl, self aligning and surface deactivated
Catalog # CRMAC4010-S630
Supplier: CHROMACOL