Order Entry
Portugal
Orders LinkContactUsLinkComponent
Titanium carbide, nanopowder

Titanium carbide, nanopowder

Glass bottle for solids, 5 g
Catalog # 44922.06
Flammable
Titanium carbide, nanopowder
Catalog # 44922.06
CAS Number:  12070-08-5
Titanium carbide, nanopowder
Titanium carbide, nanopowder

Glass bottle for solids, 5 g

CAS Number:  12070-08-5
Price
57,30 €
Log in for availability
1 * 5 g
Product Details & Documents

CAS No.: 12070-08-5 Molecular Formula: CTi Formula Weight: 59.91 Physical Form: 30-50nm APS Powder, S.A. 35-45m2/g Density: 4.93 Appearance: Black Melting Point: ca 3140° Boiling Point: 4820° UN No.: 3178 MDL No.: MFCD00011268

  • CAS Number:12070-08-5
  • Formula:CTi
  • MDL Number:MFCD00011268
  • Molecular Weight:59.89 g/mol
  • Boiling Pt:4820 °C
  • Melting Pt:∼ 3140 °C
  • Density:4.93
  • Storage Temperature:Ambient
  • EINECS:235-120-4
  • UN:3178
  • ADR:4.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications