Titanium carbide, nanopowder
Glass bottle for solids, 100 g
Supplier: Thermo Fisher Scientific
Titanium carbide, nanopowder
Catalog # 44922.22
Supplier: Thermo Fisher Scientific
CAS Number: 12070-08-5
Jump to:
Product Details & Documents
CAS No.: 12070-08-5 Molecular Formula: CTi Formula Weight: 59.91 Physical Form: 30-50nm APS Powder, S.A. 35-45m2/g Density: 4.93 Appearance: Black Melting Point: ca 3140° Boiling Point: 4820° UN No.: 3178 MDL No.: MFCD00011268
- CAS Number:12070-08-5
- Formula:CTi
- MDL Number:MFCD00011268
- Molecular Weight:59.89 g/mol
- Boiling Pt:4820 °C
- Melting Pt:∼ 3140 °C
- Density:4.93
- Storage Temperature:Ambient
- EINECS:235-120-4
- UN:3178
- ADR:4.1,III
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



