Order Entry
Portugal
ContactUsLinkComponent
Ac-asp-glu-val-asp-paranitroanilide. A colorimetric substrate for caspase-3 (km = 9.7 µ m), ccp-32 and related cysteine proteases. 1 * 10 mg
Catalog # ICNA03PNA13810
Supplier:  MP Biomedicals
Ac-asp-glu-val-asp-paranitroanilide. A colorimetric substrate for caspase-3 (km = 9.7 µ m), ccp-32 and related cysteine proteases. 1 * 10 mg
Catalog # ICNA03PNA13810
Supplier:  MP Biomedicals
Frequently Bought Together