Order Entry
Portugal
Orders LinkContactUsLinkComponent
Adapters, NS socket to spherical joint ball
Adapters, NS socket to spherical joint ball
29/32 NS, S 41
Catalog # LENZ01532941
Supplier:   Lenz Laborglas
Adapters, NS socket to spherical joint ball
Catalog # LENZ01532941
Supplier:  Lenz Laborglas
Adapters, NS socket to spherical joint ball
Adapters, NS socket to spherical joint ball
29/32 NS, S 41
Price
58,50 €
Log in for availability
1 * 1 unid.
Product Details & Documents
DURAN®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Adapters from spherical joint ball to ground joint socket.
Documents
Specifications
  • Ball:
    S 41
  • Socket:
    29/32 NS