Order Entry
Portugal
Orders LinkContactUsLinkComponent
Adapters, NS socket to spherical joint ball
Adapters, NS socket to spherical joint ball
29/32 NS, S 29
Catalog # LENZ01532929
Supplier:   Lenz Laborglas
Adapters, NS socket to spherical joint ball
Catalog # LENZ01532929
Supplier:  Lenz Laborglas
Adapters, NS socket to spherical joint ball
Adapters, NS socket to spherical joint ball
29/32 NS, S 29
Price
37,70 €
Log in for availability
1 * 1 unid.
Product Details & Documents
DURAN®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Adapters from spherical joint ball to ground joint socket.
Documents
Specifications
  • Ball:
    S 29
  • Socket:
    29/32 NS