Adapters, NS socket to spherical joint ball
14/23 NS, S 19
Catalog # LENZ01531419
Supplier: Lenz Laborglas
Jump to:
Product Details & Documents
DURAN®, borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Adapters from spherical joint ball to ground joint socket.
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
Specifications
- Ball:S 19
- Socket:14/23 NS