Order Entry
Portugal
Orders LinkContactUsLinkComponent
Anchorage lines, MH00

Anchorage lines, MH00

Anchorage line, 50 m, 14 mm
Catalog # 118-0425
Supplier:   Honeywell Safety
Anchorage lines, MH00
Catalog # 118-0425
Supplier:  Honeywell Safety
Anchorage lines, MH00
Anchorage lines, MH00

Anchorage line, 50 m, 14 mm

Price
233,00 €
Log in for availability
1 * 1 unid.
Product Details & Documents

Twisted polyamide rope suitable for 14 mm/16 mm rope grabs. Suitable for vertical, horizontal and sloping applications.

  • Reliable and lightweight
  • Breaking strength: >22 KN

Certifications: EN 353-2: Guide type fall arresters on a flexible anchorage line Comply with the standard if used with MILLER rope grab only; EN 362 (rope)

Delivery information: Supplied with a counterweight and screwgate karabiner ML00 complies with EN696

Specifications
  • Ø:
    14 mm
  • Description:
    Anchorage line
  • Length:
    50 m
  • Cat. No.:
    118-0425
  • Material:
    Polyamide