Order Entry
Portugal
Orders LinkContactUsLinkComponent
 

Anchorage lines, MH00

Anchorage line, 30 m, 14 mm
Catalog # 118-0423
Supplier:   Honeywell Safety
Anchorage lines, MH00
Catalog # 118-0423
Supplier:  Honeywell Safety
Anchorage lines, MH00
Anchorage lines, MH00

Anchorage line, 30 m, 14 mm

Price
156,00 €
Log in for availability
1 * 1 unid.
Product Details & Documents

Twisted polyamide rope suitable for 14 mm/16 mm rope grabs. Suitable for vertical, horizontal and sloping applications.

  • Reliable and lightweight
  • Breaking strength: >22 KN

 : EN 353-2: Guide type fall arresters on a flexible anchorage line Comply with the standard if used with MILLER rope grab only; EN 362 (rope)

 : Supplied with a counterweight and screwgate karabiner ML00 complies with EN696

Specifications
  • Ø:
    14 mm
  • Description:
    Anchorage line
  • Length:
    30 m
  • Cat. No.:
    118-0423
  • Material:
    Polyamide
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.