Order Entry
Portugal
Orders LinkContactUsLinkComponent
Potassium bromide ≥99%

Potassium bromide ≥99%

Glass bottle for solids, 2,5 kg
Catalog # A16339.A4
Irritant
Potassium bromide ≥99%
Catalog # A16339.A4
CAS Number:  7758-02-3
Potassium bromide ≥99%
Potassium bromide ≥99%

Glass bottle for solids, 2,5 kg

CAS Number:  7758-02-3
Price
181,00 €
Log in for availability
1 * 2,5 kg
Product Details & Documents

Purity: 99+% CAS No.: 7758-02-3 Molecular Formula: KBr Formula Weight: 119.01 Refractive Index: 1.559 Odour: Odourless Boiling Point: 1435° Merck Reference: 14,7618 MDL No.: MFCD00011358

  • CAS Number:7758-02-3
  • Formula:KBr
  • MDL Number:MFCD00011358
  • Molecular Weight:119 g/mol
  • Merck Index:14,7618
  • Boiling Pt:1435 °C
  • Melting Pt:730 °C
  • Density:2.75
  • Storage Temperature:Ambient
  • EINECS:231-830-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    2,5 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.