Order Entry
Portugal
Orders LinkContactUsLinkComponent
Dapsone 98%

Dapsone 98%

Glass bottle for solids, 500 g
Catalog # A18976.36
Irritant
Dapsone 98%
Catalog # A18976.36
CAS Number:  80-08-0
Dapsone 98%
Dapsone 98%

Glass bottle for solids, 500 g

CAS Number:  80-08-0
Price
160,00 €
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 98% CAS No.: 80-08-0 Molecular Formula: C12H12N2O2S Formula Weight: 248.30 Odour: Odourless Melting Point: 175-178° Beilstein Reference: 788055 Merck Reference: 14,2822 MDL No.: MFCD00007887

Synonyms:  4,4'-sulphonyldianiline, 4,4'-Diamino diphenyl sulphone, 4-[(4-Aminophenyl)sulphonyl]aniline, DDS, 4,4'-Sulphonyldianiline, 1-Amino-4-((4-aminophenyl)sulphonyl)benzene, 4-Aminophenyl sulphone
  • CAS Number:80-08-0
  • Formula:C₁₂H₁₂N₂O₂S
  • MDL Number:MFCD00007887
  • Molecular Weight:248.31 g/mol
  • Merck Index:14,2822
  • Melting Pt:175…178 °C
  • Storage Temperature:Ambient
  • EINECS:201-248-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.