Order Entry
Portugal
Orders LinkContactUsLinkComponent
Dapsone 98%

Dapsone 98%

Glass bottle for solids, 100 g
Catalog # A18976.22
Irritant
Dapsone 98%
Catalog # A18976.22
CAS Number:  80-08-0
Dapsone 98%
Dapsone 98%

Glass bottle for solids, 100 g

CAS Number:  80-08-0
Price
53,30 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 98% CAS No.: 80-08-0 Molecular Formula: C12H12N2O2S Formula Weight: 248.30 Odour: Odourless Melting Point: 175-178° Beilstein Reference: 788055 Merck Reference: 14,2822 MDL No.: MFCD00007887

Synonyms:  4,4'-sulphonyldianiline, 4,4'-Diamino diphenyl sulphone, 4-[(4-Aminophenyl)sulphonyl]aniline, DDS, 4,4'-Sulphonyldianiline, 1-Amino-4-((4-aminophenyl)sulphonyl)benzene, 4-Aminophenyl sulphone
  • CAS Number:80-08-0
  • Formula:C₁₂H₁₂N₂O₂S
  • MDL Number:MFCD00007887
  • Molecular Weight:248.31 g/mol
  • Merck Index:14,2822
  • Melting Pt:175…178 °C
  • Storage Temperature:Ambient
  • EINECS:201-248-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications