Azul de Timol
Glass bottle for solids, 50 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 76-61-9 Molecular Formula: C27H30O5S Formula Weight: 466.60 Melting Point: 223° dec. Merck Reference: 14,9400 MDL No.: MFCD00005869
Synonyms: Thymolsulphonphthalein, 3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
- CAS Number:76-61-9
- Formula:C₂₇H₃₀O₅S
- MDL Number:MFCD00005869
- Molecular Weight:466.6 g/mol
- Merck Index:14,9400
- Melting Pt:223 °C (decomposição)
- Storage Temperature:Ambiente
- EINECS:200-973-3
Documents
Specifications
Specifications
- Tipo de embalagem:Glass bottle for solids
- Und.:50 g



