Buffer solutions in sachets, Certipur®, Supelco®
Certipur® buffer solution, pH 12,00, potassium dihydrogen phosphate, sodium hydroxide, 12,00, Phosphate, Sachet, 30×30 ml, 30
Supplier: Merck
Jump to:
Product Details & Documents
For calibration of pH meters.
- Practical 30 ml sachets - one use per sachet - no risk of contamination
- Solutions are always fresh and ready to use
- Traceable to SRM from NIST and PTB
- Reference temperature: +25 °C
- Precision: Deviation is max. ±0,015 pH units
pH value is measured with a combined glass electrode after 5-point calibration according to DIN 19268 with reference buffer solutions according to DIN 19266, IUPAC, NIST, Ph. Eur. and USP.
Specifications
Specifications
- Pack type:Sachet
- pH buffer type:Phosphate
- Pk:30
- Description:Certipur® buffer solution, pH 12,00, potassium dihydrogen phosphate, sodium hydroxide
- Pk Info:30×30 ml
- pH value:12,00
- Cat. No.:1.99022.0001
- M-Clarity quality level:MQ100
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



