Order Entry
Portugal
Orders LinkContactUsLinkComponent
Buffer, TE solution (TRIS-EDTA buffer)

Buffer, TE solution (TRIS-EDTA buffer)

TE buffer solution (TRIS-EDTA buffer), Glass bottle, 500 mL
Catalog # PROMV6232
Supplier:   Promega
Buffer, TE solution (TRIS-EDTA buffer)
Catalog # PROMV6232
Supplier:  Promega
CAS Number:  
Buffer, TE solution (TRIS-EDTA buffer)
Buffer, TE solution (TRIS-EDTA buffer)

TE buffer solution (TRIS-EDTA buffer), Glass bottle, 500 mL

CAS Number:  
Price
326,00 €
Log in for availability
1 * 500 mL
Product Details & Documents

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Description:
    TE buffer solution (TRIS-EDTA buffer)

Specifications